Anti-SELL Rat Monoclonal Antibody [clone: MEL-14]

Lieferant: Tonbo Biosciences

70-0621-U100EA 81.5 EUR
70-0621-U100
Anti-SELL Rat Monoclonal Antibody [clone: MEL-14]
Antikörper
The MEL-14 antibody is specific for mouse CD62L, also known as L-Selectin, a cell adhesion molecule which facilitates lymphocyte “rolling” on activated vascular endothelium and homing to high endothelial venules (HEV) as immune cells transmigrate from blood into peripheral tissues. L-Selectin is a member of a family of Selectin molecules which act together with the integrin family of adhesion molecules to mediate leukocyte-endothelial interactions. L-Selectin is characteristically expressed by neutrophils, and is also found on B cells, monocytes, granulocytes, and at varying levels on naive, effector and memory T cells. It is rapidly shed upon cell activation, releasing into the circulation a soluble form whose biological role is of particular interest in cancer biology research.

Type: Primary
Antigen: SELL (selectin L)
Clonality: monoclonal
Clone: MEL-14
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR