Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]

Lieferant: Tonbo Biosciences
70-6784-U100EA 0 EUR
TNBO70-6784-U100
Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Antikörper
The 9E10 antibody was generated using a peptide (EQKLISEEDL) within the C-terminal region of human c-myc, a protein expressed in the cell nucleus, where it plays an important role in cell cycle regulation and apoptosis. In addition, c-myc is considered to be a proto-oncogenic protein as it is expressed at higher levels in a variety of hematopoietic tumors. The 9E10 antibody is widely utilized to purify or identify recombinant proteins which have been engineered to express the 9E10 epitope tag.

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: 90000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR