Anti-HA tag None Monoclonal Antibody

Lieferant: OriGene
ORIGTA100012EA 488 EUR
ORIGTA100012
Anti-HA tag None Monoclonal Antibody
Antikörper
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR