Anti-eGFP Chicken Polyclonal Antibody

Lieferant: OriGene
ORIGTA100022EA 957 EUR
ORIGTA100022
Anti-eGFP Chicken Polyclonal Antibody
Antikörper
Type: Tag
Antigen: eGFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR