Anti-DDK Tag Monoclonal Antibody (DyLight® 550)

Lieferant: OriGene
ORIGTA150022EA 468 EUR
ORIGTA150022
Anti-DDK Tag Monoclonal Antibody (DyLight® 550)
Antikörper
Type: Tag
Antigen: DDK Tag
Clonality: monoclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: None
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR