Anti-6X His tag Polyclonal Antibody

Lieferant: OriGene
ORIGTA150031EA 617 EUR
ORIGTA150031
Anti-6X His tag Polyclonal Antibody
Antikörper
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR