Anti-eGFP Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA150032EA 689 EUR
ORIGTA150032
Anti-eGFP Rabbit Polyclonal Antibody
Antikörper
Type: Tag
Antigen: eGFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR