Anti-DDK Tag Antibody Magnetic beads

Lieferant: OriGene
ORIGTA150036EA 1470 EUR
ORIGTA150036
Anti-DDK Tag Antibody Magnetic beads
Antikörper
Type: Tag
Antigen: DDK Tag
Clonality:
Clone:
Conjugation: Magnetic beads
Epitope:
Host: None
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR