Anti-tGFP Mouse Antibody (Magnetic beads)
Lieferant: OriGene
ORIGTA150040EA 1420 EUR
ORIGTA150040
Anti-tGFP Mouse Antibody (Magnetic beads)
Type: Tag
Antigen: tGFP
Clonality:
Clone:
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen