Anti-tGFP Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: OriGene
ORIGTA150043EA 583 EUR
ORIGTA150043
Anti-tGFP Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Tag
Antigen: tGFP
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR