Anti-Myc Tag None Antibody Magnetic beads

Lieferant: OriGene
ORIGTA150044EA 533 EUR
ORIGTA150044
Anti-Myc Tag None Antibody Magnetic beads
Antikörper
Type: Tag
Antigen: Myc Tag (Myc tag)
Clonality:
Clone:
Conjugation: Magnetic beads
Epitope:
Host: None
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR