Anti-His tag Monoclonal Antibody

Lieferant: OriGene
ORIGTA150088EA 510 EUR
ORIGTA150088
Anti-His tag Monoclonal Antibody
Antikörper
Type: Tag
Antigen: His tag (His tag)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR