Anti-V5 Tag Monoclonal Antibody

Lieferant: OriGene
ORIGTA150094EA 398 EUR
ORIGTA150094
Anti-V5 Tag Monoclonal Antibody
Antikörper
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR