Anti-GST tag Monoclonal Antibody

Lieferant: OriGene
ORIGTA150101EA 398 EUR
ORIGTA150101
Anti-GST tag Monoclonal Antibody
Antikörper
Type: Tag
Antigen: GST tag (Glutathione-S-Transferase tag)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR