Anti-mGFP Mouse Monoclonal Antibody

Lieferant: OriGene
ORIGTA180076EA 497 EUR
ORIGTA180076
Anti-mGFP Mouse Monoclonal Antibody
Antikörper
Type: Tag
Antigen: mGFP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR