Anti-DsRed-Monomer Mouse Monoclonal Antibody

Lieferant: OriGene
ORIGTA180083EA 398 EUR
ORIGTA180083
Anti-DsRed-Monomer Mouse Monoclonal Antibody
Antikörper
Type: Tag
Antigen: DsRed-Monomer
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR