Anti-Dendra2 Mouse Monoclonal Antibody

Lieferant: OriGene
ORIGTA180094EA 410 EUR
ORIGTA180094
Anti-Dendra2 Mouse Monoclonal Antibody
Antikörper
Type: Tag
Antigen: Dendra2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR