Anti-TEK Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300046EA 559 EUR
ORIGTA300046
Anti-TEK Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR