Anti-STK11 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300047EA 559 EUR
ORIGTA300047
Anti-STK11 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-100; Western Blot: 1:1000; Flow Cytometry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: STK11 (serine/threonine kinase 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR