Anti-PIM2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300051EA 559 EUR
ORIGTA300051
Anti-PIM2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 1:50-100; Immunochemistry: 1:50-100

Type: Primary
Antigen: PIM2 (pim-2 oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR