Anti-APP Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300060EA 698 EUR
ORIGTA300060
Anti-APP Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR