Anti-DTNBP1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300078EA 559 EUR
ORIGTA300078
Anti-DTNBP1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: DTNBP1 (dystrobrevin binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR