Anti-HDAC10 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300100EA 559 EUR
ORIGTA300100
Anti-HDAC10 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HDAC10 (histone deacetylase 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR