Anti-HS2ST1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300103EA 697 EUR
ORIGTA300103
Anti-HS2ST1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HS2ST1 (heparan sulfate 2-O-sulfotransferase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR