Anti-MEF2C Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300124EA 559 EUR
ORIGTA300124
Anti-MEF2C Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Dot Blot: 1:500

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR