Anti-SQSTM1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300177EA 559 EUR
ORIGTA300177
Anti-SQSTM1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:50-100; Western Blot: 1:1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: SQSTM1 (sequestosome 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR