Anti-CASP3 Rabbit monoclonal antibody [clone: E87]

Lieferant: OriGene
ORIGTA300366EA 675 EUR
ORIGTA300366
Anti-CASP3 Rabbit monoclonal antibody [clone: E87]
Antikörper
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:25-1:50; Flow Cytometry: 1:1000; Immunoprecipitation: 1:10

Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR