Anti-DAXX Rabbit monoclonal antibody [clone: E94]

Lieferant: OriGene
ORIGTA300370EA 685 EUR
ORIGTA300370
Anti-DAXX Rabbit monoclonal antibody [clone: E94]
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: DAXX (death-domain associated protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR