Anti-CAV1 Rabbit Monoclonal Antibody [clone: E249]

Lieferant: OriGene
ORIGTA300391EA 661 EUR
ORIGTA300391
Anti-CAV1 Rabbit Monoclonal Antibody [clone: E249]
Antikörper
Recommended Dilutions: Immunochemistry: 1:100-250; Western Blot: 1:1000-1:10000

Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR