Anti-BAD Rabbit Monoclonal Antibody [clone: Y208]

Lieferant: OriGene
ORIGTA300475EA 690 EUR
ORIGTA300475
Anti-BAD Rabbit Monoclonal Antibody [clone: Y208]
Antikörper
Recommended Dilutions: Western Blot: 1:2000-1:5000; Immunochemistry/Immunofluorescence: 1:250-1:500; Flow Cytometry: 1:20-1:50; Immunoprecipitation: 1:100

Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR