Anti-EIF5A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA300510EA 643 EUR
ORIGTA300510
Anti-EIF5A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100-1:250; Flow Cytometry: 1:20

Type: Primary
Antigen: EIF5A (eukaryotic translation initiation factor 5A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR