Anti-PTPN11 Rabbit Monoclonal Antibody [clone: Y477]
Lieferant: OriGene
ORIGTA300534EA 800 EUR
ORIGTA300534
Anti-PTPN11 Rabbit Monoclonal Antibody [clone: Y477]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunoprecipitation: 1:20
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat