Anti-EEF2K Rabbit Monoclonal Antibody [clone: EP724Y]

Lieferant: OriGene
ORIGTA300638EA 643 EUR
ORIGTA300638
Anti-EEF2K Rabbit Monoclonal Antibody [clone: EP724Y]
Antikörper
Recommended Dilutions: Western Blot: 1:50000; Immunoprecipitation: 1:20

Type: Primary
Antigen: EEF2K (eukaryotic elongation factor-2 kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR