Anti-MUC1 Rabbit Monoclonal Antibody [clone: EP1024Y]

Lieferant: OriGene
ORIGTA300711EA 661 EUR
ORIGTA300711
Anti-MUC1 Rabbit Monoclonal Antibody [clone: EP1024Y]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Flow Cytometry: 1:30; Immunoprecipitation: 1:50

Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR