Anti-AFP Rabbit Monoclonal Antibody [clone: EP1016Y]

Lieferant: OriGene
ORIGTA300729EA 643 EUR
ORIGTA300729
Anti-AFP Rabbit Monoclonal Antibody [clone: EP1016Y]
Antikörper
Recommended Dilutions: Western Blot: 1:500000; Immunoprecipitation: 1:50

Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR