Anti-CD8A Rabbit Monoclonal Antibody [clone: EP1150Y]

Lieferant: OriGene
ORIGTA300779EA 679 EUR
ORIGTA300779
Anti-CD8A Rabbit Monoclonal Antibody [clone: EP1150Y]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:50-1:500

Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR