Anti-ITGB2 Rabbit Monoclonal Antibody [clone: EP1286Y]

Lieferant: OriGene
ORIGTA300794EA 633 EUR
ORIGTA300794
Anti-ITGB2 Rabbit Monoclonal Antibody [clone: EP1286Y]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunoprecipitation: 1:50

Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR