Anti-TPH1 Rabbit Monoclonal Antibody [clone: EP1311Y]

Lieferant: OriGene
ORIGTA300818EA 661 EUR
ORIGTA300818
Anti-TPH1 Rabbit Monoclonal Antibody [clone: EP1311Y]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50 100; Flow Cytometry: 1:20; Immunoprecipitation: 1:50

Type: Primary
Antigen: TPH1 (tryptophan hydroxylase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR