Anti-NOTCH1 Rabbit Monoclonal Antibody [clone: EP1238Y]

Lieferant: OriGene
ORIGTA300836EA 694 EUR
ORIGTA300836
Anti-NOTCH1 Rabbit Monoclonal Antibody [clone: EP1238Y]
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:100; Western Blot: 1:1000-1:10000; Immunofluorescence: 1:100-1:250

Type: Primary
Antigen: NOTCH1 (notch 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR