Anti-LDHA Rabbit Monoclonal Antibody [clone: EP1563Y]
Lieferant: OriGene
ORIGTA300879EA 802 EUR
ORIGTA300879
Anti-LDHA Rabbit Monoclonal Antibody [clone: EP1563Y]
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:250; Western Blot: 1:1000-1:5000; Immunoprecipitation: 1:70; Immunochemistry: 1:100-1:250; Flow Cytometry: 1:30
Type: Primary
Antigen: LDHA (lactate dehydrogenase A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat