Anti-YAP1 Rabbit Monoclonal Antibody [clone: EP1674Y]

Lieferant: OriGene
ORIGTA300956EA 675 EUR
ORIGTA300956
Anti-YAP1 Rabbit Monoclonal Antibody [clone: EP1674Y]
Antikörper
Recommended Dilutions: Western Blot: 1:25000-1:50000; Immunoprecipitation: 1:70; Flow Cytometry: 1:50; Immunochemistry/Immunofluorescence: 1:100-1:250

Type: Primary
Antigen: YAP1 (Yes-associated protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR