Anti-RBL2 Rabbit Monoclonal Antibody [clone: EP2141Y]

Lieferant: OriGene
ORIGTA301010EA 652 EUR
ORIGTA301010
Anti-RBL2 Rabbit Monoclonal Antibody [clone: EP2141Y]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50 100; Flow Cytometry: 1:20

Type: Primary
Antigen: RBL2 (retinoblastoma-like 2 (p130))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR