Anti-RABAC1 Rabbit Monoclonal Antibody [clone: EPR1747Y]
Lieferant: OriGene
ORIGTA301019EA 763 EUR
ORIGTA301019
Anti-RABAC1 Rabbit Monoclonal Antibody [clone: EPR1747Y]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:20000; Immunoprecipitation: 1:30; Flow Cytometry: 1:100
Type: Primary
Antigen: RABAC1 (Rab acceptor 1 (prenylated))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat