Anti-NODAL Rabbit Monoclonal Antibody [clone: EP2058Y]

Lieferant: OriGene
ORIGTA301024EA 643 EUR
ORIGTA301024
Anti-NODAL Rabbit Monoclonal Antibody [clone: EP2058Y]
Antikörper
Recommended Dilutions: Flow Cytometry: 1:60; Western Blot: 1:2000

Type: Primary
Antigen: NODAL (nodal growth differentiation factor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR