Anti-YAP1 Rabbit Monoclonal Antibody [clone: EP1675Y]

Lieferant: OriGene
ORIGTA301075EA 661 EUR
ORIGTA301075
Anti-YAP1 Rabbit Monoclonal Antibody [clone: EP1675Y]
Antikörper
Recommended Dilutions: Western Blot: 1:10000-1:50000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: YAP1 (Yes-associated protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR