Anti-GRM5 Rabbit Monoclonal Antibody [clone: EPR2425Y]

Lieferant: OriGene
ORIGTA301102EA 855 EUR
ORIGTA301102
Anti-GRM5 Rabbit Monoclonal Antibody [clone: EPR2425Y]
Antikörper
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:100-250; Flow Cytometry: 1:10

Type: Primary
Antigen: GRM5 (glutamate receptor, metabotropic 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR