Anti-SOD2 Rabbit Monoclonal Antibody [clone: EPR2560Y]

Lieferant: OriGene
ORIGTA301157EA 661 EUR
ORIGTA301157
Anti-SOD2 Rabbit Monoclonal Antibody [clone: EPR2560Y]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: SOD2 (superoxide dismutase 2, mitochondrial)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR