Anti-CD5 Rabbit Monoclonal Antibody [clone: EP2952]

Lieferant: OriGene
ORIGTA301354EA 656 EUR
ORIGTA301354
Anti-CD5 Rabbit Monoclonal Antibody [clone: EP2952]
Antikörper
Recommended Dilutions: Western Blot: 1:10000-50000; Immunochemistry: 1:100-500;-250; Immunoprecipitation: 1:50

Type: Primary
Antigen: CD5 (CD5 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR