Anti-CD36 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA301401EA 526 EUR
ORIGTA301401
Anti-CD36 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:2000-1: 10000

Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR