Anti-CD36 Mouse Monoclonal Antibody [clone: D-2712]

Lieferant: OriGene
ORIGTA301402EA 526 EUR
ORIGTA301402
Anti-CD36 Mouse Monoclonal Antibody [clone: D-2712]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunoprecipitation: 1:500

Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR